Learn More
Alfa Aesar™ 1-Naphthaldehyde, 97%
Manufacturer: Alfa Aesar™ A1561018
Description
Applications1-Naphthaldehyde is used in the synthesis of single-crystalline homochiral porous metal-organic frameworks (MOFs). It is also a starting material for the Canizzaro reaction to produce 1-Naphthoic acid and 1-Naphthalenemethanol.
Solubility
Soluble in ethanol, ether, acetone, benzene. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agent, air.
Specifications
1-Naphthaldehyde | |
Pungent | |
1°C to 2°C | |
66-77-3 | |
MFCD00004003 | |
1-naphthaldehyde, 1-formylnaphthalene, 1-naphthalenecarboxaldehyde, 1-naphthylaldehyde, alpha-naphthal, alpha-naphthaldehyde, naphthaldehyde, .alpha.-naphthal, alpha-naphthylcarboxaldehyde, alpha-naphthylaldehyde | |
C1=CC=C2C(=C1)C=CC=C2C=O | |
156.184 | |
CHEBI:52367 | |
97% |
112°C (233°F) | |
160°C to 161°C (15mmHg) | |
50g | |
C11H8O | |
386082 | |
SQAINHDHICKHLX-UHFFFAOYSA-N | |
naphthalene-1-carbaldehyde | |
6195 | |
156.18 |
Chemical Identifiers
66-77-3 | |
156.184 | |
SQAINHDHICKHLX-UHFFFAOYSA-N | |
6195 | |
naphthalene-1-carbaldehyde |
C11H8O | |
MFCD00004003 | |
1-naphthaldehyde, 1-formylnaphthalene, 1-naphthalenecarboxaldehyde, 1-naphthylaldehyde, alpha-naphthal, alpha-naphthaldehyde, naphthaldehyde, .alpha.-naphthal, alpha-naphthylcarboxaldehyde, alpha-naphthylaldehyde | |
CHEBI:52367 | |
C1=CC=C2C(=C1)C=CC=C2C=O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : QJ0190000
TSCA : TSCA