Please login to your online account to display your discounted pricing

1-n-Propylpiperazine Dihydrobromide 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  161620100

 View more versions of this product

Catalog No. AC161620100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5% (on dry substance)
Formula Weight 290.04
Quantity 10g
Color Slightly yellow to light beige
EINECS Number 264-759-1
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
CAS Min % 98.5
Packaging Glass bottle
Loss on Drying 1% max. (105°C, 2 hrs)
Molecular Formula C7H16N2·2HBr
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-n-Propylpiperazine dihydrobromide
Assay Percent Range 99%
MDL Number MFCD00051992
Name Note 99%
CAS 64262-23-3

Specifications

Chemical Name or Material 1-n-Propylpiperazine dihydrobromide
Name Note 99%
CAS 64262-23-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (on dry substance)
Assay Percent Range 99%
Molecular Formula C7H16N2·2HBr
Formula Weight 290.04
MDL Number MFCD00051992
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents