Please login to your online account to display your discounted pricing

1-n-Propylpiperazine Dihydrobromide 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-n-Propylpiperazine dihydrobromide
Name Note 99%
CAS 64262-23-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (on dry substance)
Assay Percent Range 99%
Molecular Formula C7H16N2·2HBr
Formula Weight 290.04
MDL Number MFCD00051992
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC161620100

  SDS
thermo fisher scientific
161620100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5% (on dry substance)
Formula Weight 290.04
Quantity 10g
Color Slightly yellow to light beige
EINECS Number 264-759-1
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 1-n-Propylpiperazine dihydrobromide
Infrared Spectrum Authentic
Loss on Drying 1% max. (105°C, 2 hrs)
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H16N2·2HBr
MDL Number MFCD00051992
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 64262-23-3
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.