missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Propylpiperazine dihydrobromide, 98%

$66.93 - $185.05

Chemical Identifiers

CAS 64262-23-3
Molecular Formula C7H18Br2N2
Molecular Weight (g/mol) 290.043
MDL Number MFCD00051992
InChI Key YTPQLWVHCBATKO-UHFFFAOYSA-N
Synonym 1-propylpiperazine dihydrobromide, 1-n-propylpiperazine dihydrobromide, n-propylpiperazinium dibromide, n-propylpiperazine dihydrobromide, 1-n-propylpiperazine 2hbr, piperazine, 1-propyl-, dihydrobromide, acmc-209nl5, ksc494c5l, propylpiperazine, bromide, bromide, 1-n-propyl-piperazine dihydrobromide
PubChem CID 3085288
IUPAC Name 1-propylpiperazine;dihydrobromide
SMILES CCCN1CCNCC1.Br.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1025814 View Documents Alfa Aesar™
A1025814
25g Each for $66.93
 
AAA1025822 View Documents Alfa Aesar™
A1025822
100g Each for $185.05
 
Specifications

Chemical Identifiers

64262-23-3
290.043
YTPQLWVHCBATKO-UHFFFAOYSA-N
3085288
CCCN1CCNCC1.Br.Br
C7H18Br2N2
MFCD00051992
1-propylpiperazine dihydrobromide, 1-n-propylpiperazine dihydrobromide, n-propylpiperazinium dibromide, n-propylpiperazine dihydrobromide, 1-n-propylpiperazine 2hbr, piperazine, 1-propyl-, dihydrobromide, acmc-209nl5, ksc494c5l, propylpiperazine, bromide, bromide, 1-n-propyl-piperazine dihydrobromide
1-propylpiperazine;dihydrobromide

Specifications

~255°C (decomposition)
64262-23-3
MFCD00051992
1-propylpiperazine dihydrobromide, 1-n-propylpiperazine dihydrobromide, n-propylpiperazinium dibromide, n-propylpiperazine dihydrobromide, 1-n-propylpiperazine 2hbr, piperazine, 1-propyl-, dihydrobromide, acmc-209nl5, ksc494c5l, propylpiperazine, bromide, bromide, 1-n-propyl-piperazine dihydrobromide
CCCN1CCNCC1.Br.Br
290.043
290.05
1-n-Propylpiperazine dihydrobromide
25g
C7H18Br2N2
3678528
YTPQLWVHCBATKO-UHFFFAOYSA-N
1-propylpiperazine;dihydrobromide
3085288
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning