Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(n-Pentyl)cyclopentanol, 98%

$409.84 - $409.84

Chemical Identifiers

Chemical Name or Material 1-(n-Pentyl)cyclopentanol
CAS 194800-16-3
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00019284
Synonym 1-(n-Amyl)cyclopentanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2053614

  SDS
alfa aesar™
B2053614
25g Each for $409.84
Description & Specifications

Specifications

Formula Weight 156.27
CAS 194800-16-3
Chemical Name or Material 1-(n-Pentyl)cyclopentanol
Quantity 25g
Health Hazard 1 Warning
Molecular Formula C10H20O
Flash Point 82°C (180°F)
Assay 98%
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00019284
Synonym 1-(n-Amyl)cyclopentanol
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.