missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Octylpiperazine, 97%

Manufacturer:  Alfa Aesar™ H5567206

 View more versions of this product

Catalog No. AAH5567206



Specifications

Specifications

1-n-Octylpiperazine
46°C (115°F)
240°C to 241°C
5g
C12H26N2
UN1993
RRZDZQLIIMBIDL-UHFFFAOYSA-N
1-octylpiperazine
2737099
Liquid
0.836 g/mL
1.467
Colorless
54256-45-0
MFCD00958407
1-1-octyl piperazine
CCCCCCCCN1CCNCC1
198.354
198.35
97%

Chemical Identifiers

54256-45-0
198.354
RRZDZQLIIMBIDL-UHFFFAOYSA-N
2737099
CCCCCCCCN1CCNCC1
C12H26N2
MFCD00958407
1-1-octyl piperazine
1-octylpiperazine
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents