Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Hexyltheobromine, 98+%

$61.03 - $159.72

Chemical Identifiers

Chemical Name or Material 1-n-Hexyltheobromine
CAS 1028-33-7
Molecular Formula C13H20N4O2
Formula Weight 264.33
MDL Number MFCD00041424
Beilstein 270632
Merck Index 14,7127
Synonym 1-Hexyl-3,7-dimethylxanthine; Pentifylline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1359703

  SDS
alfa aesar™
L1359703
1g Each for $159.72

AAL13597MD

  SDS
alfa aesar™
L13597MD
250mg Each for $61.03
Description & Specifications

Specifications

Formula Weight 264.33
Chemical Name or Material 1-n-Hexyltheobromine
MDL Number MFCD00041424
Health Hazard 1 Warning
CAS 1028-33-7
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Assay ≥98%
Merck Index 14,7127
Synonym 1-Hexyl-3,7-dimethylxanthine; Pentifylline
Melting Point 80° to 81°C
Beilstein 270632
RTECS Number UO8450000
Quantity 1g
Molecular Formula C13H20N4O2
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.