missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Hexyltheobromine, 98+%

Manufacturer:  Alfa Aesar™ L1359703

 View more versions of this product

Catalog No. AAL1359703



Specifications

Specifications

1-n-Hexyltheobromine
1g
C13H20N4O2
270632
pentifylline, 1-hexyltheobromine, 1-n-hexyltheobromine, hexyltheobromine, cosaldon, pentifyllin, cosadon, theobromine, 1-hexyl, sk 7 pharmaceutical, 1-hexyl-3,7-dimethylxanthine
CCCCCCN1C(=O)C2=C(N=CN2C)N(C1=O)C
264.329
264.33
80°C to 81°C
1028-33-7
MFCD00041424
14,7127
MRWQRJMESRRJJB-UHFFFAOYSA-N
1-hexyl-3,7-dimethylpurine-2,6-dione
70569
≥98%

Chemical Identifiers

1028-33-7
264.329
MRWQRJMESRRJJB-UHFFFAOYSA-N
70569
CCCCCCN1C(=O)C2=C(N=CN2C)N(C1=O)C
C13H20N4O2
MFCD00041424
pentifylline, 1-hexyltheobromine, 1-n-hexyltheobromine, hexyltheobromine, cosaldon, pentifyllin, cosadon, theobromine, 1-hexyl, sk 7 pharmaceutical, 1-hexyl-3,7-dimethylxanthine
1-hexyl-3,7-dimethylpurine-2,6-dione
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UO8450000

Documents