Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Hexyl-3-methylimidazolium tetrafluoroborate, 99%

$132.38 - $628.03

Chemical Identifiers

Chemical Name or Material 1-n-Hexyl-3-methylimidazolium tetrafluoroborate
CAS 244193-50-8
Molecular Formula C10H19BF4N2
Formula Weight 254.08
MDL Number MFCD03095459
Synonym HMIMBF4
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2754806

 
alfa aesar™
H2754806
5g Each for $132.38

AAH2754818

 
alfa aesar™
H2754818
50g Each for $628.03
Description & Specifications

Specifications

Formula Weight 254.08
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium tetrafluoroborate
Assay 99%
Molecular Formula C10H19BF4N2
MDL Number MFCD03095459
CAS 244193-50-8
Synonym HMIMBF4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Melting Point −81°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.