Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide, 95%

Chemical Identifiers

Chemical Name or Material 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide
CAS 39127-10-1
Formula Weight 335.29
MDL Number MFCD00134461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5559103

 
alfa aesar™
H5559103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5559106

 
alfa aesar™
H5559106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5559114

 
alfa aesar™
H5559114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 335.29
Melting Point 125° to 128°C
Chemical Name or Material 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide
Physical Form Powder
Assay 95%
MDL Number MFCD00134461
CAS 39127-10-1
Color White to yellow
Quantity 1g