missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Decyl-3-methylimidazolium tetrafluoroborate, 97%

Manufacturer:  Alfa Aesar™ H5917906

 View more versions of this product

Catalog No. AAH5917906



Description

Description

Applications
It is employed as a intermediate for pharmaceutical.

Notes
Moisture Sensitive, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

1-n-Decyl-3-methylimidazolium tetrafluoroborate
Moisture Sensitive
5g
C14H27BF4N2
1-decyl-3-methylimidazolium tetrafluoroborate, dsstox_cid_27891, dsstox_rid_82644, dsstox_gsid_47915, ksc200g6n, c10mim bf4, 1-decyl-3-me-thylimidazolium tetrafluoroborate
[B-](F)(F)(F)F.CCCCCCCCCCN1C=C[N+](=C1)C
310.188
310.18
97%
1.4440
Yellow
244193-56-4
MFCD09038853
QGUMDWFYJYXDTH-UHFFFAOYSA-N
1-decyl-3-methylimidazol-3-ium;tetrafluoroborate
11461044
Liquid

Chemical Identifiers

244193-56-4
310.188
QGUMDWFYJYXDTH-UHFFFAOYSA-N
11461044
[B-](F)(F)(F)F.CCCCCCCCCCN1C=C[N+](=C1)C
C14H27BF4N2
MFCD09038853
1-decyl-3-methylimidazolium tetrafluoroborate, dsstox_cid_27891, dsstox_rid_82644, dsstox_gsid_47915, ksc200g6n, c10mim bf4, 1-decyl-3-me-thylimidazolium tetrafluoroborate
1-decyl-3-methylimidazol-3-ium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents