missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-n-Butylpyridinium chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01974004

 View more versions of this product

Catalog No. AA1974004



Description

Description

The derivatives of 1-n-Butylpyridinium chloride is used in spent fuel reprocessing, particularly in the extraction of high-level radioactive aqueous waste from the processing of nuclear fuel. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-n-Butylpyridinium chloride
131°C to 132°C
MFCD00060047
Hygroscopic
Soluble in water.
CCCC[N+]1=CC=CC=C1.[Cl-]
171.668
171.67
Crystalline
1124-64-7
C9H14ClN
2 g
1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
POKOASTYJWUQJG-UHFFFAOYSA-M
1-butylpyridin-1-ium;chloride
2734171
98%

Chemical Identifiers

1124-64-7
171.668
POKOASTYJWUQJG-UHFFFAOYSA-M
2734171
CCCC[N+]1=CC=CC=C1.[Cl-]
C9H14ClN
MFCD00060047
1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
1-butylpyridin-1-ium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only