missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butylpyridinium chloride, 98%

Manufacturer:  Alfa Aesar™ 1974018

 View more versions of this product

Catalog No. AA1974018



Specifications

Specifications

1-n-Butylpyridinium chloride
131°C to 132°C
1124-64-7
MFCD00060047
POKOASTYJWUQJG-UHFFFAOYSA-M
1-butylpyridin-1-ium;chloride
2734171
Crystalline
Hygroscopic
50g
C9H14ClN
1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
CCCC[N+]1=CC=CC=C1.[Cl-]
171.668
171.67
98%

Chemical Identifiers

1124-64-7
171.668
POKOASTYJWUQJG-UHFFFAOYSA-M
2734171
CCCC[N+]1=CC=CC=C1.[Cl-]
C9H14ClN
MFCD00060047
1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
1-butylpyridin-1-ium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents