Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butylpyridinium bromide, 99%

$61.64 - $293.82

Chemical Identifiers

Chemical Name or Material 1-n-Butylpyridinium bromide
CAS 874-80-6
Molecular Formula C9H14BrN
Formula Weight 216.12
MDL Number MFCD00671496
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2699506

  SDS
alfa aesar™
H2699506
5g Each for $61.64

AAH2699518

  SDS
alfa aesar™
H2699518
50g Each for $293.82
Description & Specifications

Specifications

Formula Weight 216.12
Assay 99%
Quantity 5g
MDL Number MFCD00671496
CAS 874-80-6
Melting Point 105° to 108°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H14BrN
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-n-Butylpyridinium bromide
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.