Learn More
1-(n-Butyl)imidazole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0779322
Description
1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.
Solubility
Soluble in Chloroform, Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
1-(n-Butyl)imidazole | |
0.948 | |
145°C (293°F) | |
C7H12N2 | |
MFCD00042753 | |
UN2690 | |
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl | |
MCMFEZDRQOJKMN-UHFFFAOYSA-N | |
1-butyl-1H-imidazole | |
61347 | |
99% |
4316-42-1 | |
257°C | |
Amine-like | |
1.48 | |
100 g | |
109499 | |
Soluble in Chloroform,Methanol. | |
CCCCN1C=CN=C1 | |
124.19 | |
124.19 |
Chemical Identifiers
4316-42-1 | |
124.19 | |
MCMFEZDRQOJKMN-UHFFFAOYSA-N | |
61347 | |
CCCCN1C=CN=C1 |
C7H12N2 | |
MFCD00042753 | |
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl | |
1-butyl-1H-imidazole |
Safety and Handling
GHS H Statement
H330-H318-H315-H335
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N-N-BUTYL IMIDAZOLE
EINECSNumber : 224-335-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only