Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-4-methylpyridinium tetrafluoroborate, 99%

$76.39 - $367.65

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-4-methylpyridinium tetrafluoroborate
CAS 343952-33-0
Molecular Formula C10H16BF4N
Formula Weight 237.06
MDL Number MFCD03095460
Synonym 1-n-Butyl-4-picolinium tetrafluoroborate; 4MBPBF4
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2710006

  SDS
alfa aesar™
H2710006
5g Each for $76.39

AAH2710018

  SDS
alfa aesar™
H2710018
50g Each for $367.65
Description & Specifications

Specifications

Formula Weight 237.06
Chemical Name or Material 1-n-Butyl-4-methylpyridinium tetrafluoroborate
Assay 99%
MDL Number MFCD03095460
CAS 343952-33-0
Quantity 5g
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H16BF4N
Synonym 1-n-Butyl-4-picolinium tetrafluoroborate; 4MBPBF4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.