Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-4-methylpyridinium hexafluorophosphate, 99%

$98.36 - $472.55

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-4-methylpyridinium hexafluorophosphate
CAS 401788-99-6
Molecular Formula C10H16F6NP
Formula Weight 295.22
MDL Number MFCD03095438
Synonym 1-n-Butyl-4-picolinium hexafluorophosphate; 4MBPPF6
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2735506

  SDS
alfa aesar™
H2735506
5g Each for $98.36

AAH2735518

  SDS
alfa aesar™
H2735518
50g Each for $472.55
Description & Specifications

Specifications

Formula Weight 295.22
Molecular Formula C10H16F6NP
Chemical Name or Material 1-n-Butyl-4-methylpyridinium hexafluorophosphate
Assay 99%
Melting Point 45°C
MDL Number MFCD03095438
CAS 401788-99-6
Quantity 5g
Synonym 1-n-Butyl-4-picolinium hexafluorophosphate; 4MBPPF6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.