Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-4-(ethylthio)benzene, 96%

$318.70 - $318.70

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-4-(ethylthio)benzene
CAS 216393-65-6
Molecular Formula C12H18S
Formula Weight 194.34g/mol
MDL Number MFCD01318116
Synonym 4-(Ethylthio)-n-butylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1761522

 
alfa aesar™
L17615-22
100g Each for $318.70
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material 1-n-Butyl-4-(ethylthio)benzene
CAS 216393-65-6
Assay 96%
Molecular Formula C12H18S
Formula Weight 194.34g/mol
MDL Number MFCD01318116
Synonym 4-(Ethylthio)-n-butylbenzene