Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-4-chlorobenzene, 97%

$180.09 - $534.14

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-4-chlorobenzene
CAS 15499-27-1
Molecular Formula C10H13Cl
Formula Weight 168.67
MDL Number MFCD00051926
Beilstein 1860506
Synonym 4-Chloro-n-butylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2413814

 
alfa aesar™
B2413814
25g Each for $180.09

AAB2413822

 
alfa aesar™
B2413822
100g Each for $534.14
Description & Specifications

Specifications

Formula Weight 168.67
Chemical Name or Material 1-n-Butyl-4-chlorobenzene
Assay 97%
Molecular Formula C10H13Cl
MDL Number MFCD00051926
CAS 15499-27-1
Boiling Point 225° to 228°C
Beilstein 1860506
Quantity 25g
Synonym 4-Chloro-n-butylbenzene