Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene, 99+%

$77.91 - $409.84

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene
CAS 35684-12-9
Molecular Formula C19H20O
Formula Weight 264.37
MDL Number MFCD04038796
Synonym 1-(4-n-Butylphenyl)-2-(4-methoxyphenyl)acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2730403

  SDS
alfa aesar™
H2730403
1g Each for $77.91

AAH2730409

  SDS
alfa aesar™
H2730409
10g Each for $409.84
Description & Specifications

Specifications

Formula Weight 264.37
Melting Point 40°C
Chemical Name or Material 1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene
Assay ≥99%
Molecular Formula C19H20O
MDL Number MFCD04038796
CAS 35684-12-9
Synonym 1-(4-n-Butylphenyl)-2-(4-methoxyphenyl)acetylene
Boiling Point 172° to 172°C (1mmHg)
Quantity 1g