Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene, 99+%

$58.15 - $367.65

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene
CAS 85583-83-1
Molecular Formula C20H22O
Formula Weight 278.39
MDL Number MFCD01218016
Synonym 1-(4-n-Butylphenyl)-2-(4-ethoxyphenyl)acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2710203

  SDS
alfa aesar™
H2710203
1g Each for $58.15

AAH2710209

  SDS
alfa aesar™
H2710209
10g Each for $367.65
Description & Specifications

Specifications

Melting Point 54°C
Formula Weight 278.39
Chemical Name or Material 1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene
Boiling Point 168° to 170°C (0.5mmHg)
Assay ≥99%
Quantity 1g
Molecular Formula C20H22O
MDL Number MFCD01218016
CAS 85583-83-1
Synonym 1-(4-n-Butylphenyl)-2-(4-ethoxyphenyl)acetylene