Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-3-methylpyridinium tetrafluoroborate, 99%

$81.22 - $393.27

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-3-methylpyridinium tetrafluoroborate
CAS 597581-48-1
Molecular Formula C10H16BF4N
Formula Weight 237.06
MDL Number MFCD08458499
Synonym 1-n-Butyl-3-picolinium tetrafluoroborate; 3MBPBF4
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2698306

  SDS
alfa aesar™
H2698306
5g Each for $81.22

AAH2698318

  SDS
alfa aesar™
H2698318
50g Each for $393.27
Description & Specifications

Specifications

Formula Weight 237.06
Chemical Name or Material 1-n-Butyl-3-methylpyridinium tetrafluoroborate
Assay 99%
Molecular Formula C10H16BF4N
MDL Number MFCD08458499
CAS 597581-48-1
Quantity 5g
Synonym 1-n-Butyl-3-picolinium tetrafluoroborate; 3MBPBF4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.