missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%

Manufacturer:  Alfa Aesar™ L1908718

 View more versions of this product

Catalog No. AAL1908718



Description

Description

Applications
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.

Solubility
Miscible with acetone, acetonitrile, ethyl acetate, isopropyl alcohol and methylene chloride. Immiscible with hexane, toluene and water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

1-n-Butyl-3-methylimidazolium tetrafluoroborate
174501-65-6
MFCD03095449
LSBXQLQATZTAPE-UHFFFAOYSA-N
1-butyl-3-methylimidazol-3-ium;tetrafluoroborate
2734178
≥98%
50g
C8H15BF4N2
1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate
[B-](F)(F)(F)F.CCCCN1C=C[N+](=C1)C
226.026
226.03

Chemical Identifiers

174501-65-6
226.026
LSBXQLQATZTAPE-UHFFFAOYSA-N
2734178
[B-](F)(F)(F)F.CCCCN1C=C[N+](=C1)C
C8H15BF4N2
MFCD03095449
1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate
1-butyl-3-methylimidazol-3-ium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents