missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-3-methylimidazolium nitrate, 95%

$105.49 - $988.50

Chemical Identifiers

CAS 179075-88-8
Molecular Formula C8H15N3O3
Molecular Weight (g/mol) 201.226
MDL Number MFCD08457696
InChI Key WPHIMOZSRUCGGU-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium nitrate, 1-butyl-3-methyl-1h-imidazol-3-ium nitrate, bmim no3, c4mim nitrate, ksc171c6p, 1-butyl-3-methylimidazoliumnitrate, 1-butyl-3-methyl-imidazol-3-ium nitrate, 1-butyl-3-methylimidazolium nitrate hplc
PubChem CID 16211113
IUPAC Name 1-butyl-3-methylimidazol-3-ium;nitrate
SMILES CCCCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5980306 View Documents Alfa Aesar™
H5980306
5g Each for $105.49
 
AAH5980318 View Documents Alfa Aesar™
H5980318
50g Each for $988.50
 
Specifications

Chemical Identifiers

179075-88-8
201.226
WPHIMOZSRUCGGU-UHFFFAOYSA-N
16211113
CCCCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]
C8H15N3O3
MFCD08457696
1-butyl-3-methylimidazolium nitrate, 1-butyl-3-methyl-1h-imidazol-3-ium nitrate, bmim no3, c4mim nitrate, ksc171c6p, 1-butyl-3-methylimidazoliumnitrate, 1-butyl-3-methyl-imidazol-3-ium nitrate, 1-butyl-3-methylimidazolium nitrate hplc
1-butyl-3-methylimidazol-3-ium;nitrate

Specifications

5g
C8H15N3O3
UN1479
WPHIMOZSRUCGGU-UHFFFAOYSA-N
1-butyl-3-methylimidazol-3-ium;nitrate
16211113
95%
179075-88-8
MFCD08457696
1-butyl-3-methylimidazolium nitrate, 1-butyl-3-methyl-1h-imidazol-3-ium nitrate, bmim no3, c4mim nitrate, ksc171c6p, 1-butyl-3-methylimidazoliumnitrate, 1-butyl-3-methyl-imidazol-3-ium nitrate, 1-butyl-3-methylimidazolium nitrate hplc
CCCCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]
201.226
201.22
1-n-Butyl-3-methylimidazolium nitrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.