Learn More
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1908622
Description
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
Solubility
Miscible with dichloromethane, chloroform, ethyl acetate. Immiscible with water, diethyl ether and hexane.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
1-n-Butyl-3-methylimidazolium hexafluorophosphate | |
8°C to 10°C | |
>350°C | |
1.4115 | |
100 g | |
14,1581 | |
Miscible with dichloromethane,chloroform,ethyl acetate. Immiscible with water,diethyl ether and hexane. | |
F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1 | |
284.19 | |
284.19 |
174501-64-5 | |
1.365 | |
C8H15F6N2P | |
MFCD03093295 | |
Hygroscopic | |
1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate | |
IXQYBUDWDLYNMA-UHFFFAOYSA-N | |
1-butyl-3-methylimidazol-3-ium;hexafluorophosphate | |
2734174 | |
≥98% |
Chemical Identifiers
174501-64-5 | |
284.19 | |
IXQYBUDWDLYNMA-UHFFFAOYSA-N | |
2734174 | |
F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1 |
C8H15F6N2P | |
MFCD03093295 | |
1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate | |
1-butyl-3-methylimidazol-3-ium;hexafluorophosphate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only