Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-3-methylimidazolium hexafluoroantimonate, 99%

$123.54 - $123.54

Chemical Identifiers

Chemical Name or Material 1-n-Butyl-3-methylimidazolium hexafluoroantimonate
CAS 174645-81-9
Molecular Formula C8H15F6N2Sb
Formula Weight 374.97g/mol
MDL Number MFCD07784453
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2707406

 
alfa aesar™
H27074-06
5g Each for $123.54
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 1-n-Butyl-3-methylimidazolium hexafluoroantimonate
CAS 174645-81-9
Assay 99%
Molecular Formula C8H15F6N2Sb
Formula Weight 374.97g/mol
MDL Number MFCD07784453