missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butyl-2-methyl-4-nitroimidazole, 97%

$141.47 - $486.42

Chemical Identifiers

CAS 135009-57-3
Molecular Formula C8H13N3O2
Formula Weight 183.21
MDL Number MFCD00173738
Chemical Name or Material 1-n-Butyl-2-methyl-4-nitroimidazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2489603 SDS Alfa Aesar™
B2489603
1g Each for $141.47
AAB2489606 SDS Alfa Aesar™
B2489606
5g Each for $486.42
Specifications Description & Specifications

Chemical Identifiers

135009-57-3
183.21
1-n-Butyl-2-methyl-4-nitroimidazole
C8H13N3O2
MFCD00173738

Specifications

135009-57-3
183.21
1g
97%
C8H13N3O2
MFCD00173738
1-n-Butyl-2-methyl-4-nitroimidazole