missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-n-Butoxy-2,3-difluorobenzene, 95%

Manufacturer:  Alfa Aesar™ H6191314

 View more versions of this product

Catalog No. AAH6191314



Specifications

Specifications

1-n-Butoxy-2,3-difluorobenzene
25g
C10H12F2O
1-butoxy-2,3-difluoro-benzene
CCCCOC1=C(C(=CC=C1)F)F
186.202
186.2
95%
Colorless
136239-66-2
MFCD07368738
HYDIPEHDSUNHPA-UHFFFAOYSA-N
1-butoxy-2,3-difluorobenzene
2782939
Liquid

Chemical Identifiers

136239-66-2
186.202
HYDIPEHDSUNHPA-UHFFFAOYSA-N
2782939
CCCCOC1=C(C(=CC=C1)F)F
C10H12F2O
MFCD07368738
1-butoxy-2,3-difluoro-benzene
1-butoxy-2,3-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

Documents