missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene, 90%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC426900050

 View more versions of this product

Catalog No. AC426900050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene
94.0
Authentic
Powder
90%
59° to 63°C
236.31
5g
108468-00-4
100.0
Glass bottle
≥89.0% (GC)
C13H20N2O2
NH2CH2C6H4CH2NHCO2C(CH3)3
MFCD02683058
White to slightly yellow-pink or beige

Chemical Identifiers

108468-00-4
100.0
C13H20N2O2
236.31
1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene
94.0
90%
NH2CH2C6H4CH2NHCO2C(CH3)3
MFCD02683058
≥89.0% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger