missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-N-BOC-Aminocyclopentanecarboxylic acid, 98%, Thermo Scientific™

$84.76 - $84.76

Chemical Identifiers

CAS 35264-09-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD01076126
InChI Key YBZCSKVLXBOFSL-UHFFFAOYSA-N
Synonym 1-boc-amino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, boc-cycloleucine, n-boc-cycloleucine
PubChem CID 2734645
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367340010
SDS View Documents Promotion Details
Thermo Fisher Scientific
367340010
1g Glass Bottle
Each for $84.76
 
Description

Description

Specifications

Chemical Identifiers

35264-09-6
229.28
YBZCSKVLXBOFSL-UHFFFAOYSA-N
2734645
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
C11H19NO4
MFCD01076126
1-boc-amino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, boc-cycloleucine, n-boc-cycloleucine
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid

Specifications

35264-09-6
100.0
130.0°C to 135.0°C
C11H19NO4
MFCD01076126
1g
Solubility in water: soluble in hot water. Other solubilities: soluble in methanol,insoluble in toluene
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
229.28
229.28
98%
97.5
White
98%
Authentic
Glass Bottle
1-boc-amino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, boc-cycloleucine, n-boc-cycloleucine
YBZCSKVLXBOFSL-UHFFFAOYSA-N
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
2734645
Crystalline Powder
1-N-BOC-Aminocyclopentanecarboxylic acid
SDS
Product Certifications

RUO – Research Use Only