missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-N-BOC-Aminocyclopentanecarboxylic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC367340050

 View more versions of this product

Catalog No. AC367340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-N-BOC-Aminocyclopentanecarboxylic acid
Glass bottle
130.0°C to 135.0°C
35264-09-6
100.0
C11H19NO4
1-boc-amino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, boc-cycloleucine, n-boc-cycloleucine
YBZCSKVLXBOFSL-UHFFFAOYSA-N
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
2734645
Crystalline Powder
Authentic
White
5g
97.5
98%
MFCD01076126
Solubility in water: soluble in hot water. Other solubilities: soluble in methanol,insoluble in toluene
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
229.28
229.28
98%

Chemical Identifiers

35264-09-6
229.28
YBZCSKVLXBOFSL-UHFFFAOYSA-N
2734645
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
C11H19NO4
MFCD01076126
1-boc-amino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, boc-cycloleucine, n-boc-cycloleucine
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SDS Documents