Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylthio-1-methylamino-2-nitroethylene, 97%

$29.53 - $328.47

Chemical Identifiers

Chemical Name or Material 1-Methylthio-1-methylamino-2-nitroethylene
CAS 61832-41-5
Molecular Formula C4H8N2O2S
Formula Weight 148.18
MDL Number MFCD00040994
Beilstein 2828017
Synonym N-Methyl-1-(methylthio)-2-nitroethenamine; 1-Methylmercapto-1-methylamino-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2260809

  SDS
alfa aesar™
B2260809
10g Each for $29.53

AAB2260818

  SDS
alfa aesar™
B2260818
50g Each for $78.96

AAB2260830

  SDS
alfa aesar™
B2260830
250g Each for $328.47
Description & Specifications

Specifications

Formula Weight 148.18
Quantity 10g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 61832-41-5
Melting Point 109° to 113°C
Assay 97%
Chemical Name or Material 1-Methylthio-1-methylamino-2-nitroethylene
Beilstein 2828017
Molecular Formula C4H8N2O2S
MDL Number MFCD00040994
Synonym N-Methyl-1-(methylthio)-2-nitroethenamine; 1-Methylmercapto-1-methylamino-2-nitroethylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.