missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Methylsulfonyl)imidazole, 98+%

$38.24 - $38.24

Chemical Identifiers

CAS 40736-26-3
Molecular Formula C4H6N2O2S
Molecular Weight (g/mol) 146.164
MDL Number MFCD00014496
InChI Key ZBHRCOWHFHJZFP-UHFFFAOYSA-N
Synonym n-methanesulfonylimidazole, 1-methanesulfonylimidazole, 1-methylsulfonyl-1h-imidazole, 1-methylsulphonyl-1h-imidazole, 1-mesyl-1h-imidazole, acmc-20ak2l, 1-methylsulfonyl imidazole, 1-methylsulfonyl-1h-imidazole, 1h-imidazole,1-methylsulfonyl, 1-methylsulfonyl-1h-imidazole #
PubChem CID 521009
IUPAC Name 1-methylsulfonylimidazole
SMILES CS(=O)(=O)N1C=CN=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0053106 View Documents Alfa Aesar™
L0053106
5g Each for $38.24
 
Specifications

Chemical Identifiers

40736-26-3
146.164
ZBHRCOWHFHJZFP-UHFFFAOYSA-N
521009
CS(=O)(=O)N1C=CN=C1
C4H6N2O2S
MFCD00014496
n-methanesulfonylimidazole, 1-methanesulfonylimidazole, 1-methylsulfonyl-1h-imidazole, 1-methylsulphonyl-1h-imidazole, 1-mesyl-1h-imidazole, acmc-20ak2l, 1-methylsulfonyl imidazole, 1-methylsulfonyl-1h-imidazole, 1h-imidazole,1-methylsulfonyl, 1-methylsulfonyl-1h-imidazole #
1-methylsulfonylimidazole

Specifications

86°C to 91°C
40736-26-3
MFCD00014496
n-methanesulfonylimidazole, 1-methanesulfonylimidazole, 1-methylsulfonyl-1h-imidazole, 1-methylsulphonyl-1h-imidazole, 1-mesyl-1h-imidazole, acmc-20ak2l, 1-methylsulfonyl imidazole, 1-methylsulfonyl-1h-imidazole, 1h-imidazole,1-methylsulfonyl, 1-methylsulfonyl-1h-imidazole #
CS(=O)(=O)N1C=CN=C1
146.164
146.17
1-(Methylsulfonyl)imidazole
5g
C4H6N2O2S
4177637
ZBHRCOWHFHJZFP-UHFFFAOYSA-N
1-methylsulfonylimidazole
521009
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning