Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylsulfonyl-4-nitrobenzene, 97%

$71.27 - $71.27

Chemical Identifiers

Chemical Name or Material 1-Methylsulfonyl-4-nitrobenzene
CAS 2976-30-9
Molecular Formula C7H7NO4S
Formula Weight 201.2
MDL Number MFCD00047803
Synonym 1-Methanesulfonyl-4-nitrobenzene; Methyl 4-nitrophenyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2006803

  SDS
alfa aesar™
B2006803
1g Each for $71.27
Description & Specifications

Specifications

Formula Weight 201.2
Molecular Formula C7H7NO4S
Health Hazard 1 Warning
MDL Number MFCD00047803
Melting Point 136° to 138°C
Assay 97%
Chemical Name or Material 1-Methylsulfonyl-4-nitrobenzene
CAS 2976-30-9
Synonym 1-Methanesulfonyl-4-nitrobenzene; Methyl 4-nitrophenyl sulfone
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.