missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Methylsulfonyl)-4-nitrobenzene, 95%, Maybridge

Chemical Identifiers

CAS 2976-30-9
CAS Min % 97.0
Assay Percent Range 95%
Molecular Formula C7H7NO4S
Linear Formula CH3SO2C6H4NO2
Formula Weight 201.20
MDL Number MFCD00047803
Chemical Name or Material 1-(Methylsulfonyl)-4-nitrobenzene
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RH00770DA SDS Fisher Scientific Company
RH00770DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2976-30-9
95%
CH3SO2C6H4NO2
MFCD00047803
97%
97.0
C7H7NO4S
201.20
1-(Methylsulfonyl)-4-nitrobenzene

Specifications

1g
97.0
2976-30-9
Amber Glass Bottle
95%
97%
201.20
CH3SO2C6H4NO2
MFCD00047803
C7H7NO4S
1-(Methylsulfonyl)-4-nitrobenzene