Please login to your online account to display your discounted pricing

1-(Methylsulfonyl)-4-nitrobenzene, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RH00770DA

 View more versions of this product

Catalog No. RH00770DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1g
Formula Weight 201.20
Assay 97%
CAS Min % 97.0
Linear Formula CH3SO2C6H4NO2
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 2976-30-9
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
MDL Number MFCD00047803
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 1-(Methylsulfonyl)-4-nitrobenzene
Molecular Formula C7H7NO4S
Assay Percent Range 95%

Specifications

Chemical Name or Material 1-(Methylsulfonyl)-4-nitrobenzene
CAS 2976-30-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 95%
Molecular Formula C7H7NO4S
Linear Formula CH3SO2C6H4NO2
Formula Weight 201.20
MDL Number MFCD00047803
Accessories

GHS Signal Word: Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.

SDS & Safety Documents