missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylsulfonyl-1H-benzotriazole, 97%

Manufacturer:  Alfa Aesar™ H5338614

 View more versions of this product

Catalog No. AAH5338614



Description

Description

Applications
1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

1-Methylsulfonyl-1H-benzotriazole
25g
C7H7N3O2S
1-methylsulfonyl-1h-benzotriazole, 1-methylsulfonyl-1h-benzo d 1,2,3 triazole, 1-methanesulfonyl-1h-benzotriazole, 1-methanesulfonyl-1,2,3-benzotriazole, acmc-20ap9j, cambridge id 5284919, methylsulfonyl benzotriazole, 1-methylsulfonyl benzotriazole, 1-methylsulfonyl-1h-benzotriazole, 1-methylsulphonyl-1h-benzotriazole
CS(=O)(=O)N1C2=CC=CC=C2N=N1
197.212
197.21
106°C to 108°C
37073-15-7
MFCD00595638
OMMZSDGGKVRGGP-UHFFFAOYSA-N
1-methylsulfonylbenzotriazole
792044
97%

Chemical Identifiers

37073-15-7
197.212
OMMZSDGGKVRGGP-UHFFFAOYSA-N
792044
CS(=O)(=O)N1C2=CC=CC=C2N=N1
C7H7N3O2S
MFCD00595638
1-methylsulfonyl-1h-benzotriazole, 1-methylsulfonyl-1h-benzo d 1,2,3 triazole, 1-methanesulfonyl-1h-benzotriazole, 1-methanesulfonyl-1,2,3-benzotriazole, acmc-20ap9j, cambridge id 5284919, methylsulfonyl benzotriazole, 1-methylsulfonyl benzotriazole, 1-methylsulfonyl-1h-benzotriazole, 1-methylsulphonyl-1h-benzotriazole
1-methylsulfonylbenzotriazole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement
P280-P301+P310-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents