missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylpyrrole-2-carbonyl chloride, 95%

$146.19 - $519.69

Chemical Identifiers

CAS 26214-68-6
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02094021
InChI Key JOYDZQJINHJNPM-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carbonyl chloride, 1h-pyrrole-2-carbonyl chloride, 1-methyl, 1-methyl-1h-pyrrole-2-carboxylic acid chloride, acmc-1chjw, 1-methyl-2-pyrroyl chloride, n-methyl-2-pyrroloyl chloride, n-methylpyrrolecarbonyl chloride, n-methylpyrrole carbonyl chloride, 1-methylpyrrole-2-carbonylchloride, 1-methylpyrrole-2-carboyl chloride
PubChem CID 2795092
IUPAC Name 1-methylpyrrole-2-carbonyl chloride
SMILES CN1C=CC=C1C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1843103 View Documents Alfa Aesar™
L1843103
1g Each for $146.19
 
AAL1843106 View Documents Alfa Aesar™
L1843106
5g Each for $519.69
 
Description

Description

Applications
It is applied in the preparation of 5-(Pyrrolylcarbonyl)- and 5-(Imidazolylcarbonyl)pyrimidines and use also in the novel fluorescent, minor groove and sequence specific recognition element: design, synthesis, and DNA binding properties of p-Anisylbenzimidazole-imidazole/pyrrole-containing polyamides.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store away from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Store in freezer at -20°C.
Specifications

Chemical Identifiers

26214-68-6
143.57
JOYDZQJINHJNPM-UHFFFAOYSA-N
2795092
CN1C=CC=C1C(=O)Cl
C6H6ClNO
MFCD02094021
1-methyl-1h-pyrrole-2-carbonyl chloride, 1h-pyrrole-2-carbonyl chloride, 1-methyl, 1-methyl-1h-pyrrole-2-carboxylic acid chloride, acmc-1chjw, 1-methyl-2-pyrroyl chloride, n-methyl-2-pyrroloyl chloride, n-methylpyrrolecarbonyl chloride, n-methylpyrrole carbonyl chloride, 1-methylpyrrole-2-carbonylchloride, 1-methylpyrrole-2-carboyl chloride
1-methylpyrrole-2-carbonyl chloride

Specifications

72°C to 74°C (10mmHg)
26214-68-6
MFCD02094021
472022
JOYDZQJINHJNPM-UHFFFAOYSA-N
1-methylpyrrole-2-carbonyl chloride
2795092
95%
1g
C6H6ClNO
UN3265
1-methyl-1h-pyrrole-2-carbonyl chloride, 1h-pyrrole-2-carbonyl chloride, 1-methyl, 1-methyl-1h-pyrrole-2-carboxylic acid chloride, acmc-1chjw, 1-methyl-2-pyrroyl chloride, n-methyl-2-pyrroloyl chloride, n-methylpyrrolecarbonyl chloride, n-methylpyrrole carbonyl chloride, 1-methylpyrrole-2-carbonylchloride, 1-methylpyrrole-2-carboyl chloride
CN1C=CC=C1C(=O)Cl
143.57
143.57
1-Methylpyrrole-2-carbonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : −30° to −10°C