missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylpyrrole-2-carbonitrile, TCI America™

$187.92 - $650.84

Chemical Identifiers

CAS 34884-10-1
Molecular Formula C6H6N2
Molecular Weight (g/mol) 106.128
MDL Number MFCD02693721
InChI Key JRQSGIQEBOZPHK-UHFFFAOYSA-N
Synonym 2-Cyano-1-methylpyrrole
PubChem CID 2064111
IUPAC Name 1-methylpyrrole-2-carbonitrile
SMILES CN1C=CC=C1C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27841G View Documents TCI America
M27841G
1g Each for $187.92
 
M27845G View Documents TCI America
M27845G
5g Each for $650.84
 
Specifications

Chemical Identifiers

34884-10-1
106.128
JRQSGIQEBOZPHK-UHFFFAOYSA-N
2064111
CN1C=CC=C1C#N
C6H6N2
MFCD02693721
2-Cyano-1-methylpyrrole
1-methylpyrrole-2-carbonitrile

Specifications

87°C
1g
C6H6N2
2-Cyano-1-methylpyrrole
CN1C=CC=C1C#N
106.128
106.13
≥98.0% (GC)
Yellow
34884-10-1
MFCD02693721
JRQSGIQEBOZPHK-UHFFFAOYSA-N
1-methylpyrrole-2-carbonitrile
2064111
Clear Liquid at 20°C
1-Methylpyrrole-2-carbonitrile
Safety and Handling

Safety and Handling

TSCA : No