missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylpyrazole-5-carboxylic Acid, TCI America™

$39.72 - $116.11

Chemical Identifiers

CAS 16034-46-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00464253
InChI Key JREJQAWGQCMSIY-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-5-carboxylic acid, 1-methylpyrazole-5-carboxylic acid, 2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-pyrazole-5-carboxylicacid, 5-carboxy-1-methyl-1h-pyrazole, 1-methyl-5-pyrazolecarboxylic acid, 2h-pyrazole-3-carboxylic acid, 2-methyl, acmc-209dkx, ksc173g6t
PubChem CID 643158
ChEBI CHEBI:74737
IUPAC Name 2-methylpyrazole-3-carboxylic acid
SMILES CN1C(=CC=N1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24941G View Documents TCI America
M24941G
1g Each for $39.72
 
M24945G View Documents TCI America
M24945G
5g Each for $116.11
 
Specifications

Chemical Identifiers

16034-46-1
126.115
JREJQAWGQCMSIY-UHFFFAOYSA-N
643158
2-methylpyrazole-3-carboxylic acid
C5H6N2O2
MFCD00464253
1-methyl-1h-pyrazole-5-carboxylic acid, 1-methylpyrazole-5-carboxylic acid, 2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-pyrazole-5-carboxylicacid, 5-carboxy-1-methyl-1h-pyrazole, 1-methyl-5-pyrazolecarboxylic acid, 2h-pyrazole-3-carboxylic acid, 2-methyl, acmc-209dkx, ksc173g6t
CHEBI:74737
CN1C(=CC=N1)C(=O)O

Specifications

White
1g
C5H6N2O2
1-methyl-1h-pyrazole-5-carboxylic acid, 1-methylpyrazole-5-carboxylic acid, 2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-pyrazole-5-carboxylicacid, 5-carboxy-1-methyl-1h-pyrazole, 1-methyl-5-pyrazolecarboxylic acid, 2h-pyrazole-3-carboxylic acid, 2-methyl, acmc-209dkx, ksc173g6t
CN1C(=CC=N1)C(=O)O
126.115
CHEBI:74737
Crystal-Powder at 20°C
1-Methylpyrazole-5-carboxylic Acid
225°C
16034-46-1
MFCD00464253
JREJQAWGQCMSIY-UHFFFAOYSA-N
2-methylpyrazole-3-carboxylic acid
643158
126.12
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No