missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylpyrazole-4-carboxylic Acid, TCI America™

$134.45 - $467.52

Chemical Identifiers

CAS 5952-92-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00159641
InChI Key UPPPWUOZCSMDTR-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-carboxylic acid, 1-methyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-methyl, 4-carboxy-1-methyl-1h-pyrazole, 1-methyl-pyrazole-4-carboxylic acid, 1-methylpyrazole-4-carboxylicacid, zlchem 1243, pubchem16306, pubchem20860, acmc-209me3
PubChem CID 643160
ChEBI CHEBI:74741
IUPAC Name 1-methylpyrazole-4-carboxylic acid
SMILES CN1C=C(C=N1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22245G View Documents TCI America
M22245G
5g Each for $467.52
 
M22241G View Documents TCI America
M22241G
1g Each for $134.45
 
Specifications

Chemical Identifiers

5952-92-1
126.115
UPPPWUOZCSMDTR-UHFFFAOYSA-N
643160
1-methylpyrazole-4-carboxylic acid
C5H6N2O2
MFCD00159641
1-methyl-1h-pyrazole-4-carboxylic acid, 1-methyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-methyl, 4-carboxy-1-methyl-1h-pyrazole, 1-methyl-pyrazole-4-carboxylic acid, 1-methylpyrazole-4-carboxylicacid, zlchem 1243, pubchem16306, pubchem20860, acmc-209me3
CHEBI:74741
CN1C=C(C=N1)C(=O)O

Specifications

White
5g
C5H6N2O2
1-methyl-1h-pyrazole-4-carboxylic acid, 1-methyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-methyl, 4-carboxy-1-methyl-1h-pyrazole, 1-methyl-pyrazole-4-carboxylic acid, 1-methylpyrazole-4-carboxylicacid, zlchem 1243, pubchem16306, pubchem20860, acmc-209me3
CN1C=C(C=N1)C(=O)O
126.115
CHEBI:74741
Crystal-Powder at 20°C
1-Methylpyrazole-4-carboxylic Acid
207°C
5952-92-1
MFCD00159641
UPPPWUOZCSMDTR-UHFFFAOYSA-N
1-methylpyrazole-4-carboxylic acid
643160
126.12
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No