missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylpsilocin, Tocris

Potent and selective 5-HT2C agonist

$176.10 - $741.11

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C13H18N2O
Formula Weight 218.29
Synonym 3-[2-(Dimethylamino)ethyl]-1-methylindol-4-ol
Chemical Name or Material 1-Methylpsilocin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
301710 R & D Systems
301710
10mg Each for $176.10
301750 R & D Systems
301750
50mg Each for $741.11
Specifications Description & Specifications

Chemical Identifiers

>99%
218.29
1-Methylpsilocin
C13H18N2O
3-[2-(Dimethylamino)ethyl]-1-methylindol-4-ol

Specifications

10mg
>99%
218.29
1-Methylpsilocin
1465-16-3
C13H18N2O
3-[2-(Dimethylamino)ethyl]-1-methylindol-4-ol

Safety and Handling

Recommended Storage : Store at +4°C