Please login to your online account to display your discounted pricing

1-Methylpiperidine-4-carbothioamide, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  429620050

 View more versions of this product

Catalog No. AC429620050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Color Off-white to tan
Quantity 5g
Chemical Name or Material 1-Methylpiperidine-4-carbothioamide
CAS 88654-17-5
CAS Min % 96.0
CAS Max % 100.0
Physical Form Off-white to tan powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C7H14N2S
Formula Weight 158.27g/mol
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Methylpiperidine-4-carbothioamide
CAS 88654-17-5
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C7H14N2S
Formula Weight 158.27g/mol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.