Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylpiperidine-3-methanol, 98%

$153.93 - $601.86

Chemical Identifiers

Chemical Name or Material 1-Methylpiperidine-3-methanol
CAS 7583-53-1
Molecular Formula C7H15NO
Formula Weight 129.2
MDL Number MFCD00006497
Beilstein 103713
Synonym 3-Hydroxymethyl-1-methylpiperidine; N-Methylpiperidine-3-carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2519718

 
alfa aesar™
B2519718
50g Each for $153.93

AAB2519730

 
alfa aesar™
B2519730
250g Each for $601.86
Description & Specifications

Specifications

Formula Weight 129.2
Chemical Name or Material 1-Methylpiperidine-3-methanol
Assay 98%
Molecular Formula C7H15NO
MDL Number MFCD00006497
CAS 7583-53-1
Beilstein 103713
Synonym 3-Hydroxymethyl-1-methylpiperidine; N-Methylpiperidine-3-carbinol
Health Hazard 1 Warning
Boiling Point 140° to 145°C
Quantity 50g
Flash Point 94°C (201°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.