missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylpiperazine 98.0+%, TCI America™

Manufacturer:  TCI America M0553100ML

 View more versions of this product

Catalog No. M0553100ML



Specifications

Specifications

1-Methylpiperazine
Yellow
100mL
C5H12N2
2924
PVOAHINGSUIXLS-UHFFFAOYSA-N
1-methylpiperazine
53167
Clear Liquid at 20°C
137°C
-5°C
109-01-3
MFCD00005966
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
CN1CCNCC1
100.165
100.17
≥98.0% (GC)

Chemical Identifiers

109-01-3
100.165
PVOAHINGSUIXLS-UHFFFAOYSA-N
53167
CN1CCNCC1
C5H12N2
MFCD00005966
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
1-methylpiperazine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0954

RTECSNumber : TM1225000

TSCA : Yes

Documents