Please login to your online account to display your discounted pricing

1-Methylpiperazine dihydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Methylpiperazine dihydrochloride
Name Note 98%
CAS 34352-59-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H12N2·2HCl
Formula Weight 173.09
MDL Number MFCD00012755
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC369861000

  SDS
thermo fisher scientific
AC369861000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 173.09
Quantity 100g
Color Off-white to light yellow
Melting Point 246° to 250°C
CAS Max % 100.0
Chemical Name or Material 1-Methylpiperazine dihydrochloride
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C5H12N2·2HCl
MDL Number MFCD00012755
CAS 34352-59-5
CAS Min % 97.5
Name Note 98%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.