missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylpiperazine dihydrochloride, 98%

Manufacturer:  Alfa Aesar™ H5553606

 View more versions of this product

Catalog No. AAH5553606



Specifications

Specifications

1-Methylpiperazine dihydrochloride
246°C to 250°C
34352-59-5
MFCD00012755
AILFRWRYZZVJTL-UHFFFAOYSA-N
1-methylpiperazine;dihydrochloride
2734715
Powder
White
5g
C5H14Cl2N2
1-methylpiperazine dihydrochloride
CN1CCNCC1.Cl.Cl
173.081
173.09
98%

Chemical Identifiers

34352-59-5
173.081
AILFRWRYZZVJTL-UHFFFAOYSA-N
2734715
CN1CCNCC1.Cl.Cl
C5H14Cl2N2
MFCD00012755
1-methylpiperazine dihydrochloride
1-methylpiperazine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents