Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylpentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material 1-Methylpentylzinc bromide
CAS 312693-13-3
Name Note Packaged under Argon in resealable ChemSeal™ bottles
Molecular Formula C6H13BrZn
Linear Formula CH3(CH2)3CH(CH3)ZnBr
Formula Weight 230.45
MDL Number MFCD01311409
UN Number UN3399
Synonym 2-Hexylzinc bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58543AD

  SDS
alfa aesar™
H58543AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.45
Molecular Formula C6H13BrZn
MDL Number MFCD01311409
Recommended Storage 2° to 8°C
Quantity 50mL
Synonym 2-Hexylzinc bromide
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Health Hazard 2 GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Linear Formula CH3(CH2)3CH(CH3)ZnBr
Health Hazard 1 Danger
CAS 312693-13-3
UN Number UN3399
Chemical Name or Material 1-Methylpentylzinc bromide
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Packaged under Argon in resealable ChemSeal™ bottles
Concentration 0.5M in THF
Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

2° to 8°C