missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 32387-21-6
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD01321244
InChI Key HVRCLXXJIQTXHC-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-3-carboxylic acid, 1-methyl-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid, 1-methyl, 1-methylindole-3-carboxylicacid, n-methylindole-3-carboxylic acid, pubchem7285, ramosetron impurity 7, acmc-209hsw, 1-methylindole-3-carboxilic acid
PubChem CID 854040
IUPAC Name 1-methyl-1H-indole-3-carboxylic acid
SMILES CN1C=C(C(O)=O)C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426480010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
426480010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC426480050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
426480050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

32387-21-6
175.19
HVRCLXXJIQTXHC-UHFFFAOYSA-N
854040
CN1C=C(C(O)=O)C2=CC=CC=C12
C10H9NO2
MFCD01321244
1-methyl-1h-indole-3-carboxylic acid, 1-methyl-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid, 1-methyl, 1-methylindole-3-carboxylicacid, n-methylindole-3-carboxylic acid, pubchem7285, ramosetron impurity 7, acmc-209hsw, 1-methylindole-3-carboxilic acid
1-methyl-1H-indole-3-carboxylic acid

Specifications

32387-21-6
100.0
96% min. (HPLC)
C10H9NO2
1g
HVRCLXXJIQTXHC-UHFFFAOYSA-N
1-methyl-1H-indole-3-carboxylic acid
854040
97%
96.0
Authentic
Glass bottle
MFCD01321244
1-methyl-1h-indole-3-carboxylic acid, 1-methyl-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid, 1-methyl, 1-methylindole-3-carboxylicacid, n-methylindole-3-carboxylic acid, pubchem7285, ramosetron impurity 7, acmc-209hsw, 1-methylindole-3-carboxilic acid
CN1C=C(C(O)=O)C2=CC=CC=C12
175.19
175.19
1-Methylindole-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only