missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylindole-2-carboxylic acid, ACROS Organics™

Manufacturer:  Acros Organics AC158640050

 View more versions of this product

Catalog No. AC158640050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Methylindole-2-carboxylic acid
Glass bottle
5g
C10H9NO2
22, V,3, 15
MAHAMBLNIDMREX-UHFFFAOYSA-N
1-methylindole-2-carboxylic acid
27695
Crystalline Powder
Authentic
Beige
16136-58-6
MFCD00005801
1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa
CN1C2=CC=CC=C2C=C1C(=O)O
175.187
175.19
≥97.5%

Chemical Identifiers

16136-58-6
175.187
MAHAMBLNIDMREX-UHFFFAOYSA-N
27695
CN1C2=CC=CC=C2C=C1C(=O)O
C10H9NO2
MFCD00005801
1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa
1-methylindole-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents